Learn More
Fisherbrand™ HORLOGE/BARO/THERMOMETRE DE TABLE
Innovative barometer stands on the bench
Marque: Fisherbrand™
Code nomenclature Nacres: PB.11
Informations supplémentaires : Poids : 0.01000kg
Includes: Two “AAA” alkaline batteries.
Description
Measures atmospheric pressure, graphically displays the barometric trend, and reports temperature and time. Ideal for recording ambient changes in the lab, monitoring conditions affecting sample analysis, and assisting in the prediction of weather changes.
- Graphic barometric trend shows present reading and previous 1-, 3-, 6-, and 12-hour readings in a bar-chart format
- Forecasting indicators show weather conditions for next 12 hours using symbols for sunny, cloudy, rainy, and partly cloudy
Informations sur la commande
Measures 4 x 4 x 2.25 in. (10.2 x 10.2 x 5.7cm). Weight: 4 oz.
Certifications
Traceable to standards provided by NIST™. Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA.