missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Parvalbumin Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marque: Invitrogen PA518389
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
Parvalbumin (PV) is a calcium binding protein expressed in specific muscle fibers and fast-firing neurons. PV consists of a single, unbranched chain of linked amino acids and belongs to a larger group of EF hand proteins. Studies have demonstrated that parvalbumin acts in the decay of calcium in the contraction/ relaxation cycle of fast twitch muscles. This data has shown a positive correlation between the rate of relaxation and the concentration of parvalbumin. Parvalbumin is also expressed in a specific population of GABAergic interneurones which are thought to play a role in maintaining the balance between excitation and inhibition in the cortex as well as the hippocampus. In amyotrophic lateral sclerosis (ALS) patents, parvalbumin immunoreactivity is specifically absent from neuron populations lost early in ALS.Spécification
Parvalbumin | |
Polyclonal | |
Unconjugated | |
Pvalb | |
D22S749; HGNC:9704; MGC116759; PALB1; Parv; parvalbumin; Parvalbumin (calcium binding protein); parvalbumin alpha; PV; Pva; PVALB | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
25269, 5816 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P02625, P20472 | |
Pvalb | |
Synthetic peptide sequence (GVDEFSTLVAES) corresponding to the C-terminus amino acids of PVALB | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |