Cryogenic Storage Vials
Links
Save Now - Web Exclusive Deals
Take advantage of our web exclusive prices and place your order today.
Must Have
FREESTANDING
FOR RESEARCH
USE
FOR SOLID OR LIQUID SAMPLE STORAGE
STABILITY IN a FREESTANDING POSITION
FOR HIGH RECOVERY
SOLUTIONS
FOR STORAGE OF BIOLOGICAL MATERIAL
IDENTIFY SPECIFIC VIALS QUICKLY
Complete Your Order
Complete your cryogenic storage vial order with the necessary accessories.
Great deals
Fisherbrand™ Corrugated Polypropylene Freezer Boxes
Store your frozen samples in these sturdy Fisherbrand plastic boxes. Safe to reuse multiple times.
Learn MoreGreat deals
Thermo Scientific™ Cryo Gloves
Work safely with ultra-cold materials in these mid-arm length Cryo Gloves.
Learn More
Guides to Help You Choose
Shop Your Brands
Frequently Asked Questions
Why can't I store cryogenic vials in the liquid phase of liquid nitrogen?
Cryogenic vials should only be placed in the vapor phase of liquid nitrogen. Immersion of the vials in the liquid phase could result in liquefied gas leaking into the sample. Rapid vaporization of the liquid upon removal could then possibly cause sample leakage, or even a violent explosion. Various storage tube protection options are available to prevent this.
What is the difference between externally and internally threaded cryogenic storage vials?
Externally threaded cryogenic storage vials have threads on the outer surface of the vial neck, while internally threaded vials have threads on the inner surface. The difference lies in how the cap interacts with the vial. With externally threaded vials, the cap screws onto the threads on the outside of the vial neck. This design allows for a secure seal and reduces the risk of contamination. In contrast, internally threaded vials have the threads on the inside, and the cap screws into the vial. This design offers better protection against accidental cap detachment and provides a smoother exterior surface, making them easier to handle and stack.
Can cryogenic storage vials be autoclaved?
No, cryogenic storage vials should not be autoclaved. Autoclaving involves subjecting items to high temperatures and pressure in order to sterilize them. Cryogenic storage vials are typically made of materials such as polypropylene or other plastics that are not suitable for autoclaving. The high temperatures and pressure in an autoclave can cause the vials to deform, melt, or release toxic substances. Moreover, the rubber or silicone gasket present in some vials may be damaged by autoclaving. To maintain the integrity and functionality of cryogenic storage vials, it is best to follow the manufacturer's instructions and avoid autoclaving them.