Learn More
2,3-dichloro-1-propanol, 97 %, Thermo Scientific Chemicals
Catalog Number p-3523289
Click to view available options
Quantité:
5 g
25 g
Conditionnement:
Flacon en verre
616-23-9
C3H6Cl2O
128.99
MFCD00040446
ZXCYIJGIGSDJQQ-UHFFFAOYSA-N
2,3-dichloro-1-propanol, 1-propanol, 2,3-dichloro, 2,3-dichloropropanol, alpha,beta-dichlorohydrin, 1,2-dichloro-3-propanol, glycerol alpha,beta-dichlorohydrin, 2,3-dichloropropyl alcohol, glycerol 1,2-dichlorohydrin, 2,3-dichloropropane-1-ol, ccris 954
12018
2,3-dichloropropan-1-ol
C(C(CCl)Cl)O
616-23-9
C3H6Cl2O
128.99
MFCD00040446
ZXCYIJGIGSDJQQ-UHFFFAOYSA-N
2,3-dichloro-1-propanol, 1-propanol, 2,3-dichloro, 2,3-dichloropropanol, alpha,beta-dichlorohydrin, 1,2-dichloro-3-propanol, glycerol alpha,beta-dichlorohydrin, 2,3-dichloropropyl alcohol, glycerol 1,2-dichlorohydrin, 2,3-dichloropropane-1-ol, ccris 954
12018
2,3-dichloropropan-1-ol
C(C(CCl)Cl)O
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 2, 3-Dichloro-1-propanol
% min. CAS 96.0
% max. CAS 100.0
Couleur Incolore
Densité 1.3500g/mL
Point d’ébullition 182°C
Point d’éclair 93°C
Spectre IR Authentique
Plage de pourcentage du dosage 96% min. (GC)
Conditionnement Flacon en verre
Indice de réfraction 1.4835 to 1.4855
Quantité 5 g
Gravité spécifique 1.35
Informations sur la solubilité Solubility in water: slightly soluble. Other solubilities: soluble in ether,acetone,ethanol
Poids de la formule 128.99
Pourcentage de pureté 97%
Forme physique Liquide