missing translation for 'onlineSavingsMsg'
Learn More

2,5-Acide diméthoxyphénylacétique, 99 %, Thermo Scientific Chemicals

Catalog Number p-7029818
Click to view available options
Quantité:
5 g
25 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
1758-25-4
C10H12O4
196.202
MFCD00004322
BBZDYQUXRFATHZ-UHFFFAOYSA-N
2,5-dimethoxyphenylacetic acid, 2-2,5-dimethoxyphenyl acetic acid, 2,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,5-dimethoxy, acetic acid, 2,5-dimethoxyphenyl, 2,5-dimethoxyphenyl-acetic acid, acmc-209eai, 2,5-dimethoxyphenyl, rarechem al bo 0334, 2,5-dimethoxy-phenethyl acid
74469
Acide 2-(2,5-diméthoxyphényl)acétique
COC1=CC(=C(C=C1)OC)CC(=O)O
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
1758-25-4
C10H12O4
196.202
MFCD00004322
BBZDYQUXRFATHZ-UHFFFAOYSA-N
2,5-dimethoxyphenylacetic acid, 2-2,5-dimethoxyphenyl acetic acid, 2,5-dimethoxyphenyl acetic acid, benzeneacetic acid, 2,5-dimethoxy, acetic acid, 2,5-dimethoxyphenyl, 2,5-dimethoxyphenyl-acetic acid, acmc-209eai, 2,5-dimethoxyphenyl, rarechem al bo 0334, 2,5-dimethoxy-phenethyl acid
74469
Acide 2-(2,5-diméthoxyphényl)acétique
COC1=CC(=C(C=C1)OC)CC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 2,5-Dimethoxyphenylacetic acid
Point de fusion 123°C to 125°C
Quantité 5 g
Poids de la formule 196.2
Pourcentage de pureté 99%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'