missing translation for 'onlineSavingsMsg'
Learn More

Acide 2-(carboxylméthylthio)succinique, 97 %, Thermo Scientific™

Catalog Number p-7024690
Click to view available options
Quantité:
10 g
50 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
99-68-3
C6H8O6S
208.184
MFCD00002720
JPHVSDWIWBDHOC-UHFFFAOYSA-N
carboxymethylmercaptosuccinic acid, butanedioic acid, carboxymethyl thio, carboxymethylthio succinic acid, 2-carboxymethyl sulfanyl butanedioic acid, 2-carboxymethyl thio succinic acid, carboxymethyl thio succinic acid, 2-carboxymethylsulfanyl butanedioic acid, 2-carboxymethyl sulfanyl succinic acid, butanedioic acid, 2-carboxymethyl thio
97922
Acide 2-(carboxyméthylsulfanyl) butanédioïque
C(C(C(=O)O)SCC(=O)O)C(=O)O
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
99-68-3
C6H8O6S
208.184
MFCD00002720
JPHVSDWIWBDHOC-UHFFFAOYSA-N
carboxymethylmercaptosuccinic acid, butanedioic acid, carboxymethyl thio, carboxymethylthio succinic acid, 2-carboxymethyl sulfanyl butanedioic acid, 2-carboxymethyl thio succinic acid, carboxymethyl thio succinic acid, 2-carboxymethylsulfanyl butanedioic acid, 2-carboxymethyl sulfanyl succinic acid, butanedioic acid, 2-carboxymethyl thio
97922
Acide 2-(carboxyméthylsulfanyl) butanédioïque
C(C(C(=O)O)SCC(=O)O)C(=O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 2-(Carboxymethylthio)succinic acid
Point de fusion 138°C to 141°C
Quantité 10 g
Poids de la formule 208.19
Pourcentage de pureté 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'