missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isocyanate de 2-éthylphényle, 97 %, Thermo Scientific™
Click to view available options
Quantité:
1 g
25 g
Conditionnement:
Flacon en verre ambré
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Nom chimique ou matériau | 2-Ethylphenyl isocyanate |
% min. CAS | 96.5 |
% max. CAS | 100.0 |
Densité | 1.04g/mL |
Plage de pourcentage du dosage | 97% |
Conditionnement | Flacon en verre ambré |
Formule linéaire | CH3CH2C6H4NCO |
Quantité | 1 g |
Poids de la formule | 147.18 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'