missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2H-perfluoro-5,8,11-triméthyl-3,6,9,12-tétraoxapentadécane

Marque:  Alfa Aesar™ L16953.06

Code nomenclature Nacres: NA.21

Informations supplémentaires : CAS : 26738-51-2 Poids : 0.00500kg

Quantité 5g

 Afficher plus de versions de ce produit

Code produit. 11393846

  • 68.82€ / 5g

Expédition estimée
20-05-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Spécification

Identifiants chimiques

26738-51-2
784.112
NOCLWIPRQIWFMR-UHFFFAOYSA-N
117327
C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F)(F)F)F)(F)F)F
C14HF29O4
MFCD00054717
2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-icosafluoro-5,8,11-tris trifluoromethyl-3,6,9,12-tetraoxapentadecane, 3,6,9,12-tetraoxapentadecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-eicosafluoro-5,8,11-tris trifluoromethyl, freon e 4, freon e-4, 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane, 1,1,1,2,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy-3-1,2,2,2-tetrafluoroethoxy propane
1,1,1,2,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]-3-(1,2,2,2-tétrafluoroéthoxy)propane

Spécification

2H-Perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane
None
-115°C
26738-51-2
MFCD00054717
NOCLWIPRQIWFMR-UHFFFAOYSA-N
1,1,1,2,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]-3-(1,2,2,2-tétrafluoroéthoxy)propane
117327
1.723
192°C to 195°C
5g
C14HF29O4
2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxatetradecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-icosafluoro-5,8,11-tris trifluoromethyl-3,6,9,12-tetraoxapentadecane, 3,6,9,12-tetraoxapentadecane, 1,1,1,2,4,4,5,7,7,8,10,10,11,13,13,14,14,15,15,15-eicosafluoro-5,8,11-tris trifluoromethyl, freon e 4, freon e-4, 2h-perfluoro-5,8,11-trimethyl-3,6,9,12-tetraoxapentadecane, 1,1,1,2,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,3,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propoxy propoxy-3-1,2,2,2-tetrafluoroethoxy propane
C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F)(F)F)F)(F)F)F
784.112
784.12
Vidéos
FDS Documents