Learn More
Acide 3-(difluorométhoxy)benzoïque, 97 %, Thermo Scientific™
Catalog Number p-7029200
Click to view available options
Quantité:
250 mg
1 g
4837-19-8
C8H6F2O3
188.13
MFCD00236224
OKKDGIXOKWOMRD-UHFFFAOYSA-N
3-difluoromethoxy benzoic acid, benzoic acid, 3-difluoromethoxy, 3-difluoromethoxy-benzoic acid, acmc-20anix, 3-difluoromethoxy benzoicacid, 3-carboxy-alpha,alpha-difluoroanisole
842614
Acide 3-(difluorométhoxy)benzoïque
C1=CC(=CC(=C1)OC(F)F)C(=O)O
4837-19-8
C8H6F2O3
188.13
MFCD00236224
OKKDGIXOKWOMRD-UHFFFAOYSA-N
3-difluoromethoxy benzoic acid, benzoic acid, 3-difluoromethoxy, 3-difluoromethoxy-benzoic acid, acmc-20anix, 3-difluoromethoxy benzoicacid, 3-carboxy-alpha,alpha-difluoroanisole
842614
Acide 3-(difluorométhoxy)benzoïque
C1=CC(=CC(=C1)OC(F)F)C(=O)O
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 3-(Difluoromethoxy)benzoic acid
Point de fusion 105°C
Quantité 250 mg
Poids de la formule 188.13
Pourcentage de pureté 97%