Learn More
Acide 4'-[2-(2-Pyrazinyl)-4-thiazolyl]biphényl-4-boronique, 97 %, Thermo Scientific™
Catalog Number 15433956
Click to view available options
Quantité:
1 g
5 g
2096338-84-8
C19H14BN3O2S
359.21
MFCD22682931
QIJNANKKYORYSQ-UHFFFAOYSA-N
4'-2-2-pyrazinyl-4-thiazolyl biphenyl-4-boronic acid
73996294
2096338-84-8
C19H14BN3O2S
359.21
MFCD22682931
QIJNANKKYORYSQ-UHFFFAOYSA-N
4'-2-2-pyrazinyl-4-thiazolyl biphenyl-4-boronic acid
73996294
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 4'-[2-(2-Pyrazinyl)-4-thiazolyl]biphenyl-4-boronic acid
Quantité 1 g
Poids de la formule 360.22
Pourcentage de pureté 97%