Learn More
4-(2-Fluorophényl)tétrahydropyran-4-carbonitrile, 97, +%, Thermo Scientific™
Catalog Number p-7034687
Click to view available options
Quantité:
250 mg
1 g
5 g
859164-45-7
C12H12FNO
205.232
MFCD14690885
JOYSNTZJNYCWMW-UHFFFAOYSA-N
4-2-fluorophenyl tetrahydropyran-4-carbonitrile, 4-2-fluorophenyl tetrahydro-2h-pyran-4-carbonitrile, 4-2-fluorophenyl oxane-4-carbonitrile, 4-2-fluoro-phenyl-tetrahydro-pyran-4-carbonitrile, 2h-pyran-4-carbonitrile, 4-2-fluorophenyl tetrahydro
49875771
4-(2-fluorophényl)oxane-4-carbonitrile
C1COCCC1(C#N)C2=CC=CC=C2F
859164-45-7
C12H12FNO
205.232
MFCD14690885
JOYSNTZJNYCWMW-UHFFFAOYSA-N
4-2-fluorophenyl tetrahydropyran-4-carbonitrile, 4-2-fluorophenyl tetrahydro-2h-pyran-4-carbonitrile, 4-2-fluorophenyl oxane-4-carbonitrile, 4-2-fluoro-phenyl-tetrahydro-pyran-4-carbonitrile, 2h-pyran-4-carbonitrile, 4-2-fluorophenyl tetrahydro
49875771
4-(2-fluorophényl)oxane-4-carbonitrile
C1COCCC1(C#N)C2=CC=CC=C2F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 4-(2-Fluorophenyl)tetrahydropyran-4-carbonitrile
Quantité 250 mg
Poids de la formule 205.23
Pourcentage de pureté ≥97%