Learn More
4,4’-diméthoxybenzil, 99+ %, Thermo Scientific Chemicals
Catalog Number p-114715
Click to view available options
Quantité:
5 g
25 g
Conditionnement:
Flacon en verre
1226-42-2
C16H14O4
270.27
MFCD00008405
YNANGXWUZWWFKX-UHFFFAOYSA-N
4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione
71043
1,2-bis(4-méthoxyphényl)éthane-1,2-dione
COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC
1226-42-2
C16H14O4
270.27
MFCD00008405
YNANGXWUZWWFKX-UHFFFAOYSA-N
4,4'-dimethoxybenzil, anisil, p-anisil, 1,2-bis 4-methoxyphenyl ethane-1,2-dione, bis 4-methoxyphenyl ethanedione, ethanedione, bis 4-methoxyphenyl, di-p-anisoyl, p,p'-dimethoxybenzil, 1,2-ethanedione, 1,2-bis 4-methoxyphenyl, 1,2-bis 4-methoxyphenyl-1,2-ethanedione
71043
1,2-bis(4-méthoxyphényl)éthane-1,2-dione
COC1=CC=C(C=C1)C(=O)C(=O)C2=CC=C(C=C2)OC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 4, 4'-Dimethoxybenzil
Note relative au nom 99+%
% min. CAS 99.0
% max. CAS 100.0
Point de fusion 131.0°C to 134.0°C
Couleur Jaune
Spectre IR Authentique
Plage de pourcentage du dosage 99+%
Conditionnement Flacon en verre
Formule linéaire CH3OC6H4COCOC6H4OCH3
Quantité 5 g
Beilstein 08, 428
Poids de la formule 270.27
Pourcentage de pureté 99+%
Forme physique Poudre cristalline fine