missing translation for 'onlineSavingsMsg'
Learn More

Acide 4’-éthoxybiphényle-4-boronique, 97 %, Thermo Scientific™

Catalog Number p-7052796
Click to view available options
Quantité:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
182344-29-2
C14H15BO3
242.081
MFCD06798054
ZFFXATWYKJBSCC-UHFFFAOYSA-N
4-ethoxybiphenyl-4'-boronic acid, 4'-ethoxy-1,1'-biphenyl-4-yl boronic acid, 4-ethoxybiphenyl-4-boronic acid, 4'-ethoxy-1,1'-biphenyl-4-ylboronic acid, acmc-20aixl, 4-ethoxybiphenyl-4\\'-boronic acid, 4-4-ethoxyphenyl phenyl boronic acid, 4-4-ethoxyphenyl phenylboronic acid, 4'-ethoxy-1,1'-biphenyl-4-yl boronicacid, 4'-ethoxy 1,1'-biphenyl-4-yl boronic acid
16217237
Acide [4-(4-ethoxyphenyl)phenyl]boronique
B(C1=CC=C(C=C1)C2=CC=C(C=C2)OCC)(O)O
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
182344-29-2
C14H15BO3
242.081
MFCD06798054
ZFFXATWYKJBSCC-UHFFFAOYSA-N
4-ethoxybiphenyl-4'-boronic acid, 4'-ethoxy-1,1'-biphenyl-4-yl boronic acid, 4-ethoxybiphenyl-4-boronic acid, 4'-ethoxy-1,1'-biphenyl-4-ylboronic acid, acmc-20aixl, 4-ethoxybiphenyl-4\\'-boronic acid, 4-4-ethoxyphenyl phenyl boronic acid, 4-4-ethoxyphenyl phenylboronic acid, 4'-ethoxy-1,1'-biphenyl-4-yl boronicacid, 4'-ethoxy 1,1'-biphenyl-4-yl boronic acid
16217237
Acide [4-(4-ethoxyphenyl)phenyl]boronique
B(C1=CC=C(C=C1)C2=CC=C(C=C2)OCC)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 4'-Ethoxybiphenyl-4-boronic acid
Quantité 250 mg
Poids de la formule 242.08
Pourcentage de pureté 97%
Forme physique Solide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'