Learn More
4-Méthoxyphényl 4-n-propylcyclohexanecarboxylate, 97 %, Thermo Scientific™
Catalog Number p-7036722
Click to view available options
Quantité:
1 g
5 g
67589-38-2
C17H24O3
276.376
MFCD11053431
YOMOGIXKVWWEHG-UHFFFAOYSA-N
4-methoxyphenyl 4-propylcyclohexanecarboxylate, cyclohexanecarboxylic acid, 4-propyl-, 4-methoxyphenyl ester, trans, 4-methoxyphenyl 4-propylcyclohexane-1-carboxylate, cyclohexanecarboxylic acid, 4-propyl-, 4-methoxyphenyl ester, 4-methoxyphenyl trans-4-propylcyclohexanecarboxylate, trans-4-propyl-cyclohexanecarboxylic acid 4-methoxy-phenyl ester, 4-methoxyphenyl 4-n-propylcyclohexanecarboxylate, 4-methoxyphenyl 4-propylcyclohexanecarboxylate #, trans-4-methoxyphenyl 4-propylcyclohexanecarboxylate, 4-methoxyphenyl 1s,4r-4-propylcyclohexane-1-carboxylate
106715
(4-méthoxyphényl) 4-propylcyclohexane-1-carboxylate
CCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OC
67589-38-2
C17H24O3
276.376
MFCD11053431
YOMOGIXKVWWEHG-UHFFFAOYSA-N
4-methoxyphenyl 4-propylcyclohexanecarboxylate, cyclohexanecarboxylic acid, 4-propyl-, 4-methoxyphenyl ester, trans, 4-methoxyphenyl 4-propylcyclohexane-1-carboxylate, cyclohexanecarboxylic acid, 4-propyl-, 4-methoxyphenyl ester, 4-methoxyphenyl trans-4-propylcyclohexanecarboxylate, trans-4-propyl-cyclohexanecarboxylic acid 4-methoxy-phenyl ester, 4-methoxyphenyl 4-n-propylcyclohexanecarboxylate, 4-methoxyphenyl 4-propylcyclohexanecarboxylate #, trans-4-methoxyphenyl 4-propylcyclohexanecarboxylate, 4-methoxyphenyl 1s,4r-4-propylcyclohexane-1-carboxylate
106715
(4-méthoxyphényl) 4-propylcyclohexane-1-carboxylate
CCCC1CCC(CC1)C(=O)OC2=CC=C(C=C2)OC
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 4-Methoxyphenyl 4-n-propylcyclohexanecarboxylate
Quantité 1 g
Poids de la formule 276.38
Pourcentage de pureté 97%