missing translation for 'onlineSavingsMsg'
Learn More

4-méthyl-2-(méthylthio)pyrimidine, 98 %, Thermo Scientific Chemicals

Catalog Number p-7030449
Click to view available options
Quantité:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
14001-63-9
C6H8N2S
140.204
MFCD00023242
UCERVHYBSTYCQS-UHFFFAOYSA-N
4-methyl-2-methylthio pyrimidine, 4-methyl-2 methylsulfanyl pyrimidine, 4-methyl-2-methylsulfanyl pyrimidine, pyrimidine, 4-methyl-2-methylthio, 4-methyl-2-methylsulfanyl-pyrimidine, 2-methylthio-4-methylpyrimidine, 4-methyl-2-methylthiopyrimidine, 4-methyl-2-methylsulphanyl pyrimidine, acmc-1bust, methylmethylsulfanylpyrimidine
821261
4-méthyl-2-méthylsulfanylpyrimidine
CC1=NC(=NC=C1)SC
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
14001-63-9
C6H8N2S
140.204
MFCD00023242
UCERVHYBSTYCQS-UHFFFAOYSA-N
4-methyl-2-methylthio pyrimidine, 4-methyl-2 methylsulfanyl pyrimidine, 4-methyl-2-methylsulfanyl pyrimidine, pyrimidine, 4-methyl-2-methylthio, 4-methyl-2-methylsulfanyl-pyrimidine, 2-methylthio-4-methylpyrimidine, 4-methyl-2-methylthiopyrimidine, 4-methyl-2-methylsulphanyl pyrimidine, acmc-1bust, methylmethylsulfanylpyrimidine
821261
4-méthyl-2-méthylsulfanylpyrimidine
CC1=NC(=NC=C1)SC

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 4-Methyl-2-(methylthio)pyrimidine
Densité 1.01
Point d’éclair 104°C (219°F)
Indice de réfraction 1.571
Quantité 1 g
Poids de la formule 140.21
Pourcentage de pureté 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'