missing translation for 'onlineSavingsMsg'
Learn More

4-méthyl-N-n-propylbenzamide, 97 %, Thermo Scientific™

Catalog Number p-7038223
Click to view available options
Quantité:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
39887-40-6
C11H15NO
177.25
MFCD01212045
MEKGGTFUDJLKHR-UHFFFAOYSA-N
4-methyl-n-n-propylbenzamide, benzamide, 4-methyl-n-propyl, p-methylbenzoylpropylamide, n-propyl-p-methylbenzamide
3589084
4-méthyl-N-propylbenzamide
CCCNC(=O)C1=CC=C(C)C=C1
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
39887-40-6
C11H15NO
177.25
MFCD01212045
MEKGGTFUDJLKHR-UHFFFAOYSA-N
4-methyl-n-n-propylbenzamide, benzamide, 4-methyl-n-propyl, p-methylbenzoylpropylamide, n-propyl-p-methylbenzamide
3589084
4-méthyl-N-propylbenzamide
CCCNC(=O)C1=CC=C(C)C=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 4-Methyl-N-n-propylbenzamide
Quantité 250 mg
Poids de la formule 177.25
Pourcentage de pureté 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'