Learn More
Acide 4’-(trifluorométhyl)biphényl-3-carboxylique, 97 %, Thermo Scientific™
Catalog Number 15433246
Click to view available options
Quantité:
250 mg
1 g
199528-28-4
C14H9F3O2
266.219
MFCD03424604
BJKGBAVGLRYCTK-UHFFFAOYSA-N
4'-trifluoromethyl-biphenyl-3-carboxylic acid, 4'-trifluoromethyl-1,1'-biphenyl-3-carboxylic acid, 3-4-trifluoromethyl phenyl benzoic acid, 4'-trifluoromethylbiphenyl-3-carboxylic acid, 4'-trifluoromethyl 1,1'-biphenyl-3-carboxylic acid, 4'-trifluoromethyl biphenyl-3-carboxylic acid, 1,1'-biphenyl-3-carboxylicacid, 4'-trifluoromethyl, pubchem9318, amtda080
2782718
Acide 3-[4-(trifluorométhyl)phényl]benzoïque
C1=CC(=CC(=C1)C(=O)O)C2=CC=C(C=C2)C(F)(F)F
199528-28-4
C14H9F3O2
266.219
MFCD03424604
BJKGBAVGLRYCTK-UHFFFAOYSA-N
4'-trifluoromethyl-biphenyl-3-carboxylic acid, 4'-trifluoromethyl-1,1'-biphenyl-3-carboxylic acid, 3-4-trifluoromethyl phenyl benzoic acid, 4'-trifluoromethylbiphenyl-3-carboxylic acid, 4'-trifluoromethyl 1,1'-biphenyl-3-carboxylic acid, 4'-trifluoromethyl biphenyl-3-carboxylic acid, 1,1'-biphenyl-3-carboxylicacid, 4'-trifluoromethyl, pubchem9318, amtda080
2782718
Acide 3-[4-(trifluorométhyl)phényl]benzoïque
C1=CC(=CC(=C1)C(=O)O)C2=CC=C(C=C2)C(F)(F)F
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 4'-(Trifluoromethyl)biphenyl-3-carboxylic acid
Quantité 1 g
Poids de la formule 266.22
Pourcentage de pureté 97%