missing translation for 'onlineSavingsMsg'
Learn More

Sel de cyclohexylammonium 6-chloro-3-indolyl-bêta-D-glucuronide, 98 %, Thermo Scientific™

Catalog Number p-7025537
Click to view available options
Quantité:
10 mg
50 mg
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
138182-20-4
C14H13ClNO7
342.71
MFCD00153940
UFBPRKNLSYGHJJ-BYNIDDHOSA-M
cyclohexanaminium 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxytetrahydro-2h-pyran-2-carboxylate, salmon-glca, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate, c14h14clno7.c6h13n, 6-chloro-3-indolyl b-d-glucuronide cyclohexyl, 6-chloro-3-indolyl beta-d-glucuronide cyclohexylammonium salt, 6-chloro-3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt
16211694
Acide (2S,3S,4S,5R,6S)-6-[(6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylique ;cyclohexanamine
O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC(Cl)=CC=C23)O[C@@H]([C@H]1O)C([O-])=O
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
138182-20-4
C14H13ClNO7
342.71
MFCD00153940
UFBPRKNLSYGHJJ-BYNIDDHOSA-M
cyclohexanaminium 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxytetrahydro-2h-pyran-2-carboxylate, salmon-glca, cyclohexylammonium ion 2s,3s,4s,5r,6s-6-6-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate, c14h14clno7.c6h13n, 6-chloro-3-indolyl b-d-glucuronide cyclohexyl, 6-chloro-3-indolyl beta-d-glucuronide cyclohexylammonium salt, 6-chloro-3-indoxyl-beta-d-glucuronic acid, cyclohexylammonium salt
16211694
Acide (2S,3S,4S,5R,6S)-6-[(6-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylique ;cyclohexanamine
O[C@@H]1[C@@H](O)[C@H](OC2=CNC3=CC(Cl)=CC=C23)O[C@@H]([C@H]1O)C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau 6-Chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt
Quantité 10 mg
Sensibilité Light and moisture sensitive
Poids de la formule 442.9
Pourcentage de pureté 98%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'