missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Kit ELISA pour estradiol
Immunoessai enzymatique pour la mesure quantitative d’estradiol libre actif dans la salive
sur devis
Spécification
Méthode de détection | Colorimetric |
---|---|
Type de produit | Estradiol ELISA Kit |
Contenu et stockage | Store the kit at 4°C |
Includes:
- Wash Concentrate concentrate for 1200mL, 1 x 30mL
- Substrate ready to use, Tetramethylbenzidine (TMB), 1 x 25mL
- Stop Solution ready to use, containing 0.5M H2SO4, 1 x 14mL
- Standard A, ready to use, 0pg/mL,1 x 1mL
- Standard B ready to use, 1pg/mL, 1 x 1mL
- Standard C ready to use, 5pg/mL, 1 x 1mL
- Standard D ready to use, 10pg/mL,1 x 1mL
- Standard E ready to use, 50pg/mL,1 x 1mL
- Standard F ready to use, 100pg/mL, 1 x 1mL
- Microtiter wells 12 x 8 (break apart) strips, 96 wells; Wells coated with an anti-Estradiol antibody (polyclonal), 1 x 96 wells
- Enzyme Conjugate ready to use, 1 x 26mL
- Control 1 ready to use, 1 x 1mL
- Control 2 ready to use, 1 x 1mL
Spécification
Colorimetric | |
Estradiol ELISA Kit | |
Store the kit at 4°C |