missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | ACAP2 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16812504
|
Proteintech
14029-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16802504
|
Proteintech
14029-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The low density lipoprotein (LDL) receptor system coordinates the metabolism of cholesterol, an essential component of the plasma membrane of all mammalian cells. Study of this system has led to an enhanced understanding of the cellular basis of cholesterol homeostasis. It has also brought into focus an important mechanism of metabolic regulation-the process of receptor-mediated endocytosis. Data suggest that the juxtamembranous region of the cytoplasmic domain participates in protein:protein interactions that allow the low density lipoprotein receptor to cluster in coated pits. It has been shown that the family of LDL receptors may serve as viral receptors. Endocytosis of the Flaviviridae viruses, hepatitis C virus, GB virus C/hepatitis G virus, and bovine viral diarrheal virus (BVDV) was shown to be mediated by LDL receptors on cultured cells.Spécification
ACAP2 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q15057, Q5FVC7, Q6ZQK5 | |
23527, 619382, 78618 | |
ACAP2 Fusion Protein Ag5190 | |
Primary | |
-20°C | |
ACAP2 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ACAP2, Centaurin beta 2, CENTB2, CNT B2, KIAA0041 | |
Acap2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |