missing translation for 'onlineSavingsMsg'
Learn More

ACAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14029-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802504

  • 329.85€ / 150µL

Date d’expédition estimée: 18-06-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The low density lipoprotein (LDL) receptor system coordinates the metabolism of cholesterol, an essential component of the plasma membrane of all mammalian cells. Study of this system has led to an enhanced understanding of the cellular basis of cholesterol homeostasis. It has also brought into focus an important mechanism of metabolic regulation-the process of receptor-mediated endocytosis. Data suggest that the juxtamembranous region of the cytoplasmic domain participates in protein:protein interactions that allow the low density lipoprotein receptor to cluster in coated pits. It has been shown that the family of LDL receptors may serve as viral receptors. Endocytosis of the Flaviviridae viruses, hepatitis C virus, GB virus C/hepatitis G virus, and bovine viral diarrheal virus (BVDV) was shown to be mediated by LDL receptors on cultured cells.
Spécification

Spécification

ACAP2
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q15057, Q5FVC7, Q6ZQK5
ACAP2, Centaurin beta 2, CENTB2, CNT B2, KIAA0041
Rabbit
IgG
150 μL
-20°C
Primary
23527, 619382, 78618
Immunoprecipitation, Western Blot
Unconjugated
ACAP2
Liquid
Acap2
ACAP2 Fusion Protein Ag5190
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents