missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems ActiveMax™ Human IL-4 Protein, Tag Free

215.94€ - 479.87€

Spécification

Numéro d’adhésion AAH67514
Poids moléculaire 15.0 kDa
Source ActiveMax™ Human IL-4, Tag Free (IL4-H4218) is expressed from human 293 cells (HEK293). It contains AA His 25 - Ser 153 (Accession # P05112-1).
Immunogène His 25 - Ser 153
Conditions de stockage -20°C
Consulter d'autres spécifications

Produits 2
Code produit Marque Quantité Prix Quantité et disponibilité  
Code produit Marque Quantité Prix Quantité et disponibilité  
15877047
Afficher les documents Détails de la promotion
ACROBiosystems
IL4-H4218-50UG
50 μg
479.87€
50µg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 10-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
15867047
Afficher les documents Détails de la promotion
ACROBiosystems
IL4-H4218-20UG
20 μg
215.94€
20µg
Voir les produits de remplacement
Économisez jusqu'à 
»
Expédition estimée: 10-04-2024
pour voir le stock disponible 
Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!
Description

Description

Interleukin-4, is a cytokine that induces differentiation of naive helper T cells (Th0 cells to Th2 cells). In the presence of IL-4 and IL-13, cytokines that are produced in a Th-2 type response, particularly during allergy and parasitic infections, macrophages become differentially activated, And this cytokine is a ligand for interleukin 4 receptor. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play a central role in mediating the immune regulatory signal of this cytokine. Recently, researcher found that the cytokine IL-4 plays a key role in development of innate CD8+ T cells in the thymus of several gene-deficient mouse strains, including Itk, KLF2, CBP and Id3, without previous exposure to antigen.

This protein carries no "e;tag"e;. The protein has a calculated MW of 15.0 kDa. The protein migrates as 15 kDa and 19 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

AAH67514
ActiveMax™ Human IL-4, Tag Free (IL4-H4218) is expressed from human 293 cells (HEK293). It contains AA His 25 - Ser 153 (Accession # P05112-1).
-20°C
IL-4
Human
PBS, pH7.4
HEK293
ActiveMax™ Cytokines
15.0 kDa
His 25 - Ser 153
IL4,BCGF1,BSF1
Unconjugated
Tag Free
Less than 0.1 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
Vidéos
FDS
Documents

Documents

Product Certifications