missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACYP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13433-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Acylphosphatase is a small cytosolic enzyme that catalyzes the hydrolysis of the carboxyl-phosphate bond of acylphosphates. Two isoenzymes have been isolated, called muscle acylphosphatase and erythrocyte acylphosphatase, on the basis of their tissue localization. This gene encodes the erythrocyte acylphosphatase isoenzyme. Alternatively spliced transcript variants that encode different proteins were identified through data analysis.Spécification
ACYP1 | |
0.58 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07311, P56376 | |
Acylphosphatase 1, ACYP1, ACYPE | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
299203, 66204, 97 |
Western Blot | |
Unconjugated | |
ACYP1 | |
Liquid | |
Acyp1 | |
ACYP1 Fusion Protein Ag4237 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Rat, Mouse |