missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26928-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
ADAM12 is a member of the ADAM (a disintegrin and metalloprotease-like domain) family. Two forms of ADAM12 have been described: ADAM12S and ADAM12L (short and long forms). The short form is a soluble form lacking the transmembrane and cytoplasmic domains. The short form of ADAM12 was reported to provoke myogenesis, and the lack of cytoplasmic domain suggests different regulation pathways (although both forms can be expressed in the same tissue).Spécification
ADAM12 | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43184, Q61824 | |
ADAM 12, ADAM12, MCMP, MCMPMltna, Meltrin alpha, MLTN, MLTNA | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
11489, 679837, 8038 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
ADAM12 | |
Liquid | |
Adam12 | |
ADAM12 Fusion Protein Ag17474 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |