missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13706-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
GTPase-activating protein for the ADP ribosylation factor family (Potential). Binds phosphatidylinositol 3, 4, 5-trisphosphate (PtdInsP3) and inositol 1, 3, 4, 5-tetrakisphosphate (InsP4). Possesses a stoichiometry of two binding sites for InsP4 with identical affinity.Spécification
ADAP2 | |
Polyclonal | |
Unconjugated | |
ADAP2 | |
ADAP2, ArfGAP with dual PH domains 2, cent b, CENTA2, Centaurin alpha 2, Cnt a2, HSA272195 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
216991, 55803, 56826 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8R2V5, Q9JK15, Q9NPF8 | |
ADAP2 | |
ADAP2 Fusion Protein Ag4641 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |