missing translation for 'onlineSavingsMsg'
Learn More

AFG3L2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 14631-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16803374

  • 112.71€ / 20µL

Expédition estimée
03-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

AFG3L2 is a protein localized in mitochondria and closely related to paraplegin. The paraplegin gene is responsible for an autosomal recessive form of hereditary spastic paraplegia. AFG3L2 gene is a candidate gene for other hereditary spastic paraplegias or neurodegenerative disorders.This gene encodes a protein localized in mitochondria and closely related to paraplegin. The paraplegin gene is responsible for an autosomal recessive form of hereditary spastic paraplegia. This gene is a candidate gene for other hereditary spastic paraplegias or neurodegenerative disorders.
Spécification

Spécification

AFG3L2
0.36 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8JZQ2, Q9Y4W6
AFG3 like protein 2, AFG3L2, Paraplegin like protein
Rabbit
IgG
20 μL
-20°C
Primary
10939, 307350, 69597
Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot
Unconjugated
AFG3L2
Liquid
Afg3l2
AFG3L2 Fusion Protein Ag6209
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Rat, Mouse
Vidéos
FDS
Documents