missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AKD1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23042-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
AKD1 | |
Polyclonal | |
Unconjugated | |
AK9 | |
AKD1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
221264 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5TCS8 | |
AK9 | |
AKD1 Fusion Protein Ag19038 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |