missing translation for 'onlineSavingsMsg'
Learn More

AKD2 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 27554-1-AP-20UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16805695

  • 112.71€ / 20µL

Expédition estimée
25-02-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Involved in maintaining the homeostasis of cellular nucleotides by catalyzing the interconversion of nucleoside phosphates. Has both nucleoside monophosphate and diphosphate kinase activities. Catalyzes the phosphorylation of AMP, dAMP, CMP and dCMP with ATP as phosphate donor and of CMP with GTP as phosphate donor. Also catalyzes the production of ATP, CTP, GTP, UTP, dATP, dCTP, dGTP and TTP from the corresponding diphosphate substrates with either ATP or GTP as phosphate donor. Shows substrate preference of CDP > UDP > ADP > GDP > TDP.
Spécification

Spécification

AKD2
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q5TCS8
Adenylate kinase 9, AK 9, AK9, AKD1, AKD2, C6orf199, C6orf224, RP1 70A9.1
Rabbit
IgG
20 μL
-20°C
Primary
221264
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
AK9
Liquid
AK9
AKD2 Fusion Protein Ag25805
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents