missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EnquireBio AKT1 (Prognostic Marker for Neuroendocrine Tumors) Mouse, Clone: AKT1/2552, enQuireBio™
Mouse Monoclonal Antibody
Marque: EnquireBio 207-MSM2-P1
Informations supplémentaires : Poids : 0.01000kg
Spécification
AKT1 (Prognostic Marker for Neuroendocrine Tumors) | |
Nuclear, Cytoplasmic and Cell Surface | |
AKT1/2552 | |
Unconjugated | |
Mouse | |
62kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
207 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant fragment of human AKT1 protein (apprx. aa 85-189) | |
PDGF-treated NIH/3T3 cells. HeLa cell lysates. Human pancreas or cervical carcinoma. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |