Learn More
MP Biomedicals™ Albumin, Human, Fraction V Protease, HRP, and AP Free, 10g
Marque: MP Biomedicals™ 08823002
Code nomenclature Nacres: NA.28
Informations supplémentaires : CAS : 70024-90-7 Poids : 0.01000kg
Spécification
1mg/mL | |
Human Albumin Fraction V | |
10g | |
Powder |
Immunoassay, Mammalian Cell Culture | |
6.8 | |
2° to 8°C |