Learn More
ALDH5A1 Mouse anti-Human, Unconjugated, Clone: 1B9H10, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67347-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The ALDH5A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ALDH5A1. This antibody recognizes human, rat, mouse, pig antigen. The ALDH5A1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
ALDH5A1 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse, Rat, Pig |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1B9H10 | |
Unconjugated | |
20 μL | |
7915 | |
IgG1 |