Learn More
ALDH5A1 Mouse anti-Human, Unconjugated, Clone: 1B9H10, Proteintech
Mouse Monoclonal Antibody
Marque:  Proteintech 67347-1-IG-20UL
Informations supplémentaires : Poids : 0.00010kg
Code produit. 16829935
  • 140.00€ / 20µL
Expédition estimée: 06-05-2024
Description
The ALDH5A1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ALDH5A1. This antibody recognizes human, rat, mouse, pig antigen. The ALDH5A1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Spécification
Spécification
ALDH5A1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
1B9H10
Unconjugated
20 μL
7915
IgG1