Learn More
ALS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Thermo Scientific PA559252
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.02000kg
Description
Immunogen sequence: DCGCPLKALR DFALQNPSAV PRFVQAICEG DDCQPPAYTY NNITCASPPE VVGLDLRDLS EAHFAP Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 77%.
The protein encoded by this gene is a serum protein that binds insulin-like growth factors, increasing their half-life and their vascular localization. Production of the encoded protein, which contains twenty leucine-rich repeats, is stimulated by growth hormone. Defects in this gene are a cause of acid-labile subunit deficiency, which maifests itself in a delayed and slow puberty. Three transcript variants encoding two different isoforms have been found for this gene.Spécification
ALS | |
Polyclonal | |
Unconjugated | |
IGFALS | |
ACLSD; Albs; Als; IGFALS; insulin like growth factor binding protein acid labile subunit; insulin-like growth factor binding protein complex acid labile chain; insulin-like growth factor binding protein complex acid-labile subunit; insulin-like growth factor binding protein, acid labile subunit; insulin-like growth factor-binding protein complex acid labile chain; insulin-like growth factor-binding protein complex acid labile subunit; mKIAA4111 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3483 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P35858 | |
IGFALS | |
Recombinant protein corresponding to Human ALS | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only