Learn More
Oxyde d’aluminium, pour la décoloration, Thermo Scientific™
Catalog Number p-7080040
Click to view available options
Quantité:
250 g
1 kg
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
1344-28-1
Al2O3
101.96
MFCD00003424
PNEYBMLMFCGWSK-UHFFFAOYSA-N
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
Pour la décoloration
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Point d’ébullition 2980°C
Nom chimique ou matériau Aluminum oxide
Point de fusion 2045°C
Couleur Blanc
Forme physique Poudre
Quantité 250 g
Formule linéaire Al2O3
Merck Index 14,356
Informations sur la solubilité Insoluble in water.
Nom IUPAC trioxidandiide de dialuminium(3+)
Poids de la formule 101.96
Odeur Odorless
Indice de réfraction 1.765