missing translation for 'onlineSavingsMsg'
Learn More

Amylase alpha Mouse anti-Human, Porcine, Rat, Clone: 2F1G8, Proteintech

Mouse Monoclonal Antibody

Marque:  Proteintech 66133-1-IG-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16827735

  • 399.00€ / 150µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This antibody can recognize amylase alpha.

Amylases are secreted proteins that hydrolyze 1,4-alpha-glucoside bonds in oligosaccharides and polysaccharides, and thus catalyze the first step in digestion of dietary starch and glycogen. The human genome has a cluster of several amylase genes that are expressed at high levels in either salivary gland or pancreas. It is an amylase isoenzyme produced by the salivary gland.
Spécification

Spécification

Amylase alpha
Monoclonal
0.79 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04746
AMY2A, Amy2a3
Amylase, alpha Fusion Protein Ag8819
150 μL
Primary
Human, Porcine, Rat
Antibody
IgG2b
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
2F1G8
Unconjugated
AMY2A
AMY2A, Amylase, alpha, Pancreatic alpha amylase
Mouse
Protein A
RUO
100521789, 279, 497039
-20°C
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions