missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANGPTL8/Betatrophin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23792-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
The Betatrophin protein, also known as the hepatocellular carcinoma-associated protein TD26, is an adipocyte- and liver-enriched insulin targeted gene that is thought to play a role in lipid metabolism be regulating serum triglyceride levels. Betatrophin is closely related to the angiopoietin-like (ANGPTL) protein family. Betatrophin mRNA increases in the cold environment and after treatment with a high-fat diet, while its mRNA levels decrease in response to fasting, suggesting that it may play a role in mediating thermogenesis and energy homeostasis.Spécification
ANGPTL8/Betatrophin | |
Polyclonal | |
Unconjugated | |
ANGPTL8 | |
ANGPTL8, Betatrophin, C19orf80, LOC55908, PRO1185, PVPA599, td26 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55908, 624219 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6UXH0, Q8R1L8 | |
Angptl8 | |
ANGPTL8/Betatrophin Fusion Protein Ag20698 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |