missing translation for 'onlineSavingsMsg'
Learn More

ATP Synthase α Mouse, Unlabeled, Clone: 51, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 612516

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15876199

  • 266.00€ / 1 pièce
Date d’expédition estimée: 17-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

ATP synthase is a large enzyme complex that uses an electrochemical H+ or Na+ gradient to synthesize ATP from ADP and Pi, providing the organism with the ATP needed for energy. The complex consists of two major units, F0 and F1. F0 is embedded in the inner membrane of the mitochondria and, due to its hydrophobic nature, translocates protons across this membrane. F1 is the catalytic portion in the matrix region of the mitochondria and is comprised of α, β, γ, δ, and ε subunits at a 3:3:1:1:1 ratio. The α subunit is a ubiquitous protein that is highly conserved among species. It has an adenine specific binding site that binds both ATP and ADP. There are two glycine rich regions in the sequence, the A domain and B domain, that are thought to be part of the nucleotide binding domain. It has been demonstrated that the N-terminus of the α subunit is necessary for the correct functional and structural connection of F0 to F1. Thus, the α subunit is an essential component of the ATP synthase complex and plays a role in properly orienting the F0 and F1 units.

Immunofluorescence, Western Blotting

Spécification

Spécification

ATP Synthase α
51
Unconjugated
Affinity Purified
Human ATP Synthase α aa. 113-220
50μg
Cell Biology
Primary
Canine, Chicken, Human, Murine, Rat
Western Blot
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.