Learn More
Casein Kinase IIα/α Mouse, Unlabeled, Clone: 31, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 611610
Informations supplémentaires : Poids : 0.01100kg
Description
Casein kinase II (CKII, CK2) is a pleiotropic, ubiquitous, and constitutively active Ser/Thr kinase that utilizes both ATP and GTP as phosphoryl donors to catalyze the phosphorylation of numerous protein substrates. It has been implicated in DNA replication, control of metabolism, and regulation of transcription and translation. CKII is a messenger independent kinase due to the fact that its activity is not affected by factors that regulate other protein kinases (e.g. cAMP, Ca2+, diacylglycerol). It is composed of two catalytic (α and α′) subunits and two regulatory (β) subunits. The β subunits stabilize the holoenzyme and enhance the activity of the α subunits. Although the enzyme exists as a tetramer, the free α/α′ subunits are catalytically active by themselves and are present in cells under some conditions. The α and α′ subunits (85% homologous) are encoded by distinct genes and have been detected in multiple species including human, bovine, and avian. CKII is found primarily in the nucleus in dividing cells, but can be detected in the cytoplasm in quiescent cells. Thus, CKII activity is required for cell cycle progression and other essential signal transduction events.
Host Species: Mouse
Clone: 31
Isotype: IgG1
Species Reactivity [for Features Main]: Rat
Immunogen: Rat Casein Kinase II α aa. 1-123
Immunofluorescence, Western Blotting
Spécification
Casein Kinase IIα/α | |
Monoclonal | |
250μg/mL | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Rat Casein Kinase II α aa. 1-123 | |
50 μg | |
Cell Biology | |
Rat, Mouse, Human | |
IgG1 |
Western Blot | |
31 | |
Unconjugated | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.