Learn More
CD107b Mouse anti-Human, FITC, Clone: H4B4, BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 555804
Informations supplémentaires : Poids : 0.02000kg
Description
Reacts with the heavily glycosylated 120kDa lysosmal-associated protein, LAMP-2. LAMP-2 is a widely expressed intracellular antigen. It is also expressed on the surface of activated platelets, PHA-activated lymphocytes and some tumor cell lines, including U937 and KG1a. LAMP-2 has been shown to be a ligand for E-selectin-mediated cell adhesion. LAMP-1 (CD107a) and LAMP-2 are carriers for poly-N-acetyllactosamines, and are able to display sialyl Le[x] termini.
Host Species: Mouse
Clone: H4B4
Isotype: IgG1 κ
Species Reactivity: Human
Intracellular Staining
Spécification
CD107b | |
Monoclonal | |
FITC | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Human Adult Adherent Peripheral Blood Cells | |
100 Tests | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
H4B4 | |
LAMP-2 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.