Learn More
SEMA7A Monoclonal Antibody (MEM-150), FITC, Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA119589
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
CD108 (Sema7A) is a GPI-anchored semaphorin family member, which enhances central and peripheral axonal growth and is required for proper axon track formation during ebryogenesis. CD108 also regulates osteoclast differentiation and pre-osteoblastic cell migration, and in immune system affects cell proliferation, chemotaxis and cytokine release. On erythrocytes CD108 defines the JMH (John-Milton-Hagen) human blood group. CD108 signalizes through its receptors - plexin C1 and beta1 integrins.
SEMA7A Monoclonal antibody specifically detects SEMA7A in Human samples. It is validated for Flow Cytometry.Spécification
SEMA7A | |
Monoclonal | |
FITC | |
SEMA7A | |
2900057C09Rik; CD108; CDw108; H-Sema K1; H-SEMA-K1; H-Sema-L; JMH; JMH blood group antigen; John Milton Hagen blood group H-Sema K1; john-Milton-Hargen human blood group Ag; MGC126692; MGC126696; M-Sema-L; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A; sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, (semaphorin) 7A (JMH blood group); sema domain, immunoglobulin domain (Ig), and GPI membrane anchor, 7A; sema K1; Sema L; SEMA7A; SEMAK1; SEMAL; semaphorin 7A (John Milton Hagen blood group); semaphorin 7A, GPI membrane anchor; semaphorin 7A, GPI membrane anchor (John Milton Hagen blood group); Semaphorin K1; semaphorin-7A; Semaphorin-K1; semaphorin-L; Semk1 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
8482 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
MEM-150 | |
TBS with 0.2% BSA and 15mM sodium azide; pH 8.0 | |
O75326 | |
SEMA7A | |
T cell line HPB-ALL. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgM |