missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Kit Monoclonal Antibody (ACK4), Invitrogen™
Rat Monoclonal Antibody
Marque: Invitrogen MA170079
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.01000kg
Description
c-Kit Monoclonal antibody specifically detects c-Kit in Mouse samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot.Spécification
c-Kit | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P05532 | |
KIT | |
IL-3 dependent mast cells derived from WB- +/+ mice | |
250 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
ACK4 | |
Unconjugated | |
KIT | |
belly-spot; Bs; CD117; ckit; C-Kit; c-KIT gene1; c-kit protein; c-kit proto-oncogene protein; c-kit receptor; c-kit receptor tyrosine kinase; dominant spotting; Dominant white spotting; Fdc; Gsfsco1; Gsfsco5; Gsfsow3; KIT; kit oncogene; KIT proto-oncogene receptor tyrosine kinase; KIT proto-oncogene, receptor tyrosine kinase; mast cell growth factor receptor; mast/stem cell growth factor receptor; mast/stem cell growth factor receptor Kit; MGF; p145 c-kit; PBT; Piebald trait protein; protein kinase; proto-oncogene c-Kit; proto-oncogene tyrosine-protein kinase Kit; receptor tyrosine kinase c-kit; receptor tyrosine kinase type III; SCFR; SCO1; SCO5; Sl; soluble KIT variant 1; SOW3; spotted sterile male; Ssm; Steel Factor Receptor; Tr-kit; tyrosine kinase receptor; tyrosine kinase receptor c-KIT; tyrosine-protein kinase Kit; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog; v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein; W | |
Rat | |
Protein G | |
RUO | |
16590 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |