Learn More
CD11c Mouse anti-Human, BB515, Clone: B-ly6, BD
Mouse Monoclonal Antibody
221.00€ - 467.00€
Spécification
Antigène | CD11c |
---|---|
Clone | B-ly6 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugué | Brilliant Blue 515 (BB515) |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15873679
|
BD Biosciences
564491 |
25 Tests |
221.00€
1 pièce |
Expédition estimée: 22-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15863679
|
BD Biosciences
564490 |
100 Tests |
467.00€
1 pièce |
Expédition estimée: 22-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The B-ly6 monoclonal antibody specifically binds to the 150 kDa adhesion glycoprotein CD11c (p150, integrin α chain). CD11c is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. It associates with CD18 to form the CD11c/CD18 complex that binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 plays a role as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium.
The antibody was conjugated to BD Horizon™ BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
Spécification
CD11c | |
Flow Cytometry | |
Brilliant Blue 515 (BB515) | |
RUO | |
Spleen Cells from Human with Hairy Cell Leukemia | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
B-ly6 | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified | |
ITGAX; AlphaX integrin chain; Axb2; Integrin alpha-X; CR4; Leu M5; SLEB6 |
For Research Use Only.