missing translation for 'onlineSavingsMsg'
Learn More

CD11c Mouse anti-Human, PE-Cy5, Clone: B-LY6, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 561692

Informations supplémentaires : Poids : 0.02000kg

 Afficher plus de versions de ce produit

Code produit. 15871239

  • 199.92€ / 1 pièce
Date d’expédition estimée: 28-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The B-ly6 monoclonal antibody specifically binds to the 150 kDa adhesion glycoprotein CD11c (p150, integrin α chain). CD11c is expressed on dendritic cells, monocytes, macrophages, granulocytes, NK cells and subsets of B and T cells. It associates with CD18 to form the CD11c/CD18 complex that binds fibrinogen and has been reported to be a receptor for iC3b and ICAM-1. Reports indicate that CD11c/CD18 plays a role as an adhesion molecule that mediates cellular binding to ligands expressed on stimulated epithelium and endothelium.

Spécification

Spécification

CD11c
B-LY6
ITGAX; AlphaX integrin chain; Axb2; Integrin alpha-X; CR4; Leu M5; SLEB6
Affinity Purified
Spleen Cells from Human with Hairy Cell Leukemia
25 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PE-Cyanine5
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Primary
Human
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.