missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, Brilliant Violet 786, Clone: M5E2, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 563699

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15873009

  • 265.20€ / 1 pièce
Date d’expédition estimée: 23-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The M5E2 monoclonal antibody specifically binds to CD14, a 53–55 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein expressed at high levels on monocytes. Additionally, the anti-CD14 antibody reacts with interfollicular macrophages, reticular dendritic cells, and some Langerhans cells. CD14 has been identified as a high affinity cell-surface receptor for complexes of lipopolysaccharide (LPS) and serum LPS-binding protein, LPB.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon™ BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).

Spécification

Spécification

CD14
M5E2
LPS receptor; LPS-R; Myeloid cell-specific leucine-rich glycoprotein
Affinity Purified
Human CD14 Protein
25 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Brilliant Violet 786
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG2a, κ
RUO
Primary
Baboon, Canine, Cynomolgus, Human, Rhesus Monkey
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.