missing translation for 'onlineSavingsMsg'
Learn More

CD20 Mouse anti-Human, PE-CF594, Clone: 2H7, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 562295

Informations supplémentaires : Poids : 0.00840kg

 Afficher plus de versions de ce produit

Code produit. 15871789

  • 545.00€ / 1 pièce
Date d’expédition estimée: 14-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The 2H7 monoclonal antibody specifically binds to CD20 that is encoded by the MS4A1 (Membrane-spanning 4-domains, subfamily A, member 1) gene. CD20 is a 33-37 kDa unglycosylated four-transmembrane phosphoprotein. CD20 is expressed on pre-B-cells, resting and activated B cells and follicular dendritic cells but not on plasma cells. Low level CD20 expression is observed on a small subset of normal circulating T lymphocytes. The CD20 molecule is involved in the regulation of B-cell activation.

Spécification

Spécification

CD20
2H7
MS4A1; B1; Bp35; LEU-16; S7
Affinity Purified
Human 6.16c1.3 B cell line
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
PE-CF594
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG2b, κ
RUO
Primary
Human
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.