missing translation for 'onlineSavingsMsg'
Learn More

CD200 Mouse anti-Human, PE, Clone: MRC OX-104, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 561762

Informations supplémentaires : Poids : 0.02000kg

 Afficher plus de versions de ce produit

Code produit. 15871309

  • 251.00€ / 1 pièce
Expédition estimée: 15-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The MRC OX-104 monoclonal antibody specifically binds to CD200. CD200 is a 40-45 kDa type 1 transmembrane glycoprotein and is also referred to as OX2. CD200 is a member of the immunoglobulin superfamily of proteins. It contains two Ig domains, a single transmembrane region and a short cytoplasmic tail. CD200 is expressed on resting and PMA-activated T- and B-cells, but not on NK cells, monocytes, granulocytes, or platelets. It has been found on a subset of CD34+ progenitor cells. Interaction of CD200 with its receptor on macrophages induces a downregulation of macrophage activity in a variety of tissues, suggesting a regulatory function.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Spécification

Spécification

CD200
Monoclonal
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human CD200 Recombinant Protein
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
MRC OX-104
OX-2; MOX1; MOX2 ; My033
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.