Learn More
CD235a Mouse anti-Human, APC, Clone: GA-R2 (HIR2), BD
Mouse Monoclonal Antibody
Marque: BD Biosciences 551336
Informations supplémentaires : Poids : 0.01000kg
Description
The GA-R2 (also known as HIR2) monoclonal antibody specifically binds to CD235a, otherwise known as Glycophorin A (GYPA, GPA), PAS-2, MN sialoglycoprotein (MNS, MN), and Sialoglycoprotein A/alpha. CD235a is a type I transmembrane sialoglycoprotein that is expressed on human erythrocytes, erythroid precursor cells and certain leukemic cell types. This antibody is useful for the identification and characterization of erythrocytes, certain myeloid leukemic cell types, and studies of erythroid cell development and infectious diseases with erythrocyte involvement. Mature, non-nucleated red blood cells characteristically express very high levels of glycophorin A.
Spécification
CD235a | |
Monoclonal | |
0.2mg/mL | |
Glycophorin-A; GYPA; GPA; PAS-2; MN sialoglycoprotein; Sialoglycoprotein A | |
Mouse | |
0.1 mg | |
Immunology | |
Human | |
IgG2b κ |
Flow Cytometry | |
GA-R2 (HIR2) | |
APC | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.