missing translation for 'onlineSavingsMsg'
Learn More

BCAM Monoclonal Antibody (BRIC221), Invitrogen™

Mouse Monoclonal Antibody

Marque:  Invitrogen MA516591

334.68 EUR valable jusqu'au 2024-03-29
Utilisez le code promo "21615" pour bénéficier de cette offre.



Code nomenclature Nacres: NA.41

Informations supplémentaires : Poids : 0.01000kg

Code produit. 15235869

  • 502.00€ / 200µg
Expédition estimée: 22-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Non-reducing conditions are recommended for Western blot applications. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL.

BCAM Monoclonal antibody specifically detects BCAM in Human samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot.
Spécification

Spécification

BCAM
Monoclonal
1 mg/mL
tris with glycine and <0.1% sodium azide
P50895
BCAM
Human erythrocytes
200 μg
Primary
Human
Antibody
IgG2b
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot
BRIC221
Unconjugated
BCAM
1200005K12Rik; antigen identified by monoclonal antibody F8; AU; Auberger B antigen; basal cell adhesion molecule; basal cell adhesion molecule (Lu and Au blood groups); basal cell adhesion molecule (Lutheran blood group); basal cell adhesion molecule Lutheran blood group; BCAM; B-CAM; B-CAM cell surface glycoprotein; B-cell adhesion molecule; CD239; F8/G253 antigen; glycoprotein 95kDa; Gplu; LU; lutheran antigen; Lutheran blood group; Lutheran blood group (Auberger b antigen included); Lutheran blood group glycoprotein; Lutheran blood group variant LUGA; lutheran glycoprotein; MSK19
Mouse
Protein G
RUO
4059
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions