missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, APC, Clone: HIT3A, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 555342

Informations supplémentaires : Poids : 0.02000kg

 Afficher plus de versions de ce produit

Code produit. 15868078

  • 457.00€ / 1 pièce
Expédition estimée: 10-05-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The HIT3a monoclonal antibody specifically binds to the human CD3 ε-chain, a 20 kDa subunit of the CD3/T cell antigen receptor complex found on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody can be mitogenic for T lymphocytes. The CD3 complex plays a role in signal transduction during antigen recognition by the T cell receptor for antigen. HIT3a antibody does not stain intracellular CD3 unlike the other CD3-specific clone, UCHT1.

Spécification

Spécification

CD3
Monoclonal
APC
Mouse
100 Tests
Immunology
Human
IgG2a κ
Flow Cytometry
HIT3A
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions

For Research Use Only.