missing translation for 'onlineSavingsMsg'
Learn More

CD3 Mouse anti-Human, APC-Cy7, Clone: SK7, BD

Mouse Monoclonal Antibody

Marque:  BD Biosciences 557832

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15869068

  • 535.50€ / 1 pièce
Date d’expédition estimée: 17-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

The SK7 (Leu-4) monoclonal antibody specifically binds to the epsilon chain of the CD3 antigen/T-cell antigen receptor (TCR) complex. This complex is composed of at least six proteins that range in molecular weight from 20 to 30 kDa. The antigen recognized by CD3 antibodies is noncovalently associated with either α/β or γ/δ TCR (70 to 90 kDa). The CD3 antigen is present on 61% to 85% of normal peripheral blood lymphocytes 60% to 85% of thymocytes and on Purkinje cells in cerebellum. The soluble form of this antibody has a mitogenic effect on most peripheral blood T lymphocytes, provided appropriate functional monocytes are present.

Spécification

Spécification

CD3
SK7
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
APC-Cyanine7
Affinity Purified
Human Thymocytes
100 Tests
Immunology
Primary
Human
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

For Research Use Only.