Learn More
CD307e (FcRL5) Monoclonal Antibody (307307), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA524020
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.00010kg
Description
In direct ELISAs, 100% cross-reactivity with recombinant human (rh) FCRL3 is observed and no cross-reactivity with rhFCRL1, 2, or 4 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
CD307e (FcRL5) Monoclonal antibody specifically detects CD307e (FcRL5) in Human samples. It is validated for Flow Cytometry.Spécification
CD307e (FcRL5) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q96RD9 | |
FCRL5 | |
Mouse myeloma cell line NS0-derived recombinant human FCRL5/FCRL3 Gln16-Arg844 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry | |
307307 | |
Unconjugated | |
FCRL5 | |
BXMAS1; BXMAS1-like protein 2; CD307; CD307e; fc receptor homolog 3; Fc receptor homolog 5; Fc receptor like 5; Fc receptor-like 3; Fc receptor-like 5; Fc receptor-like protein 3; Fc receptor-like protein 5; Fcrh3; FCRH5; Fcrl3; Fcrl5; FcR-like protein 5; Immune receptor translocation-associated protein 2; immunoglobulin superfamily receptor translocation associated 2 (IRTA2); IRTA2; mBXMH2; moFcRH3; PRO820; rCG62831-like; UNQ503/PRO820 | |
Mouse | |
Protein A/G | |
RUO | |
83416 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |